4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(2,2,2-trifluoroethoxy)pyridine 97%
Supplier: Ambeed
|
Warning
|
A323720-100MG
A323720-250MG
103825-356EA
469
USD
103825-356
77263-478
77263-480
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(2,2,2-trifluoroethoxy)pyridine 97%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(2,2,2-trifluoroethoxy)pyridine
Precaución: for Research Use Only
Formula:
C₁₃H₁₇BF₃NO₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD29117191 CAS Number: 1415748-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...