1H-Pyrazole-4-boronic acid pinacol ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
A311208-1G
A311208-25G
A311208-5G
A311208-100G
A311208-10G
A311208-500G
103814-656EA
5
USD
103814-656
103835-844
76838-632
76838-634
76838-636
77250-972
77250-974
77250-976
1H-Pyrazole-4-boronic acid pinacol ester 98%
1H-Pyrazole-4-boronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₉H₁₅BN₂O₂ Melting Pt: 142…146 °C Flash Pt: 156.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD03453063 CAS Number: 269410-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...