4-Indoleboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

A194704-10G A194704-250MG A194704-25G A194704-5G A194704-1G
103816-686EA 10 USD
103816-686 103847-298 76769-958 76769-960 77085-222 77085-224 77302-432
4-Indoleboronic acid pinacol ester 97%
4-Indoleboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BNO₂
MW: 243.11 g/mol
Boiling Pt: 396 °C (760 mm Hg)
Melting Pt: 120…125 °C
Flash Pt: 193.3 °C
Storage Temperature: Ambient
MDL Number: MFCD08689896
CAS Number: 388116-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR