4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-pyrazole 95%

Supplier: Ambeed
Warning

A212835-100MG A212835-250MG A212835-1G A212835-5G
103818-244EA 122 USD
103818-244 103848-940 76986-780 76986-782 77087-418 77087-420
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-pyrazole 95%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₅H₂₉BN₂O₃Si
MW: 324.29 g/mol
Boiling Pt: 395.1±27.0 °C (760 mm Hg)
Flash Pt: 192.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD20486100
CAS Number: 894807-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR