1H-Indazole-4-boronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-1H-indazole, 1H-indazol-4-boronic acid pinacol ester

A186744-100MG A186744-10G A186744-250MG A186744-1G A186744-5G A186744-25G
103815-958EA 90 USD
103815-958 103846-546 76985-930 76985-932 76985-934 77084-228 77084-230 77890-163
1H-Indazole-4-boronic acid pinacol ester 97%
1H-Indazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BN₂O₂
MW: 244.10 g/mol
Boiling Pt: 404.1 °C (760 mm Hg)
Melting Pt: 139…142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08059267
CAS Number: 885618-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR