4-(4-(Trifluoromethyl)phenyl)oxazol-2-amine 96%

Supplier: Ambeed
Warning

A286111-1G A286111-100MG
103813-702EA 139 USD
103813-702 103834-838 76837-368 77032-220
4-(4-(Trifluoromethyl)phenyl)oxazol-2-amine 96%
4-(4-(Trifluoromethyl)phenyl)oxazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₇F₃N₂O
MW: 228.171 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14706776
CAS Number: 859721-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR