4-(4-(tert-Butyl)phenyl)-1H-pyrazol-3-amine 97%

Supplier: Ambeed
Warning

A121009-250MG A121009-1G
103800-020EA 93 USD
103800-020 77009-436 77312-132
4-(4-(tert-Butyl)phenyl)-1H-pyrazol-3-amine 97%
4-(4-(tert-Butyl)phenyl)-1H-pyrazol-3-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₇N₃
MW: 215.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06634630
CAS Number: 1015845-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR