4-(4-(Methylsulfonyl)phenyl)piperidine 95%

Supplier: Ambeed
Warning

A557953-1G A557953-250MG
103828-732EA 111 USD
103828-732 76843-966 76991-742
4-(4-(Methylsulfonyl)phenyl)piperidine 95%
4-(4-(Methylsulfonyl)phenyl)piperidine

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₂S
Storage Temperature: Ambient
MDL Number: MFCD04114999
CAS Number: 885274-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR