N-(3-Hydroxypropyl)morpholine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Morpholinopropanol, 3-Morpholino-1-propanol, 4-(3-Hydroxypropyl)morpholine, 3-(4-Morpholinyl)-1-propanol

A136573-10G A136573-1G
103802-322EA 6 USD
103802-322 77109-574 77315-232
N-(3-Hydroxypropyl)morpholine 98%
N-(3-Hydroxypropyl)morpholine

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂
MW: 145.2 g/mol
Boiling Pt: 240…241 °C (760 mm Hg)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD01713533
CAS Number: 4441-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR