4-(3-Hydroxypropyl)benzonitrile 95%

Supplier: Ambeed
Warning

A141066-1G A141066-5G
103802-944EA 123 USD
103802-944 77299-190 77299-192
4-(3-Hydroxypropyl)benzonitrile 95%
4-(3-Hydroxypropyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO
Storage Temperature: Refrigerator
MDL Number: MFCD09028771
CAS Number: 83101-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR