4-(3-(Trimethoxysilyl)propyl)morpholine 98%
Supplier: Ambeed
|
Warning
|
A101261-10G
A101261-1G
A101261-5G
103795-878EA
10
USD
103795-878
103821-562
77148-200
77297-028
77297-030
4-(3-(Trimethoxysilyl)propyl)morpholine 98%
4-(3-(Trimethoxysilyl)propyl)morpholine
Precaución: for Research Use Only
Formula:
C₁₀H₂₃NO₄Si MW: 249.38 g/mol Boiling Pt: 266 °C (760 mm Hg) Flash Pt: 115 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00053579 CAS Number: 31024-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...