4-(2-Methoxyethoxy)cyclohexanol 95%
Supplier: Ambeed
|
Warning
|
A577414-100MG
A577414-250MG
103828-842EA
193
USD
103828-842
103839-992
76649-926
76649-928
4-(2-Methoxyethoxy)cyclohexanol 95%
4-(2-Methoxyethoxy)cyclohexanol
Precaución: for Research Use Only
Formula:
C₉H₁₈O₃ MW: 174.23 g/mol Storage Temperature: Ambient |
CAS Number:
107025-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...