4-(2-Hydroxyethyl)oxindole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(2-Hydroxyethyl)indolin-2-one, 4-(2-Hydroxyethyl)-2-indolinone
A210973-100G
A210973-10G
A210973-250MG
A210973-1G
A210973-25G
A210973-5G
103818-060EA
14
USD
103818-060
103848-752
76975-548
76975-550
76975-552
77087-138
77087-140
77303-164
4-(2-Hydroxyethyl)oxindole 98%
4-(2-Hydroxyethyl)oxindole
Precaución: for Research Use Only
Formula:
C₁₀H₁₁NO₂ MW: 177.20 g/mol Boiling Pt: 390 °C (760 mm Hg) Melting Pt: 148…150 °C Flash Pt: 189.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD07782133 CAS Number: 139122-19-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...