4-(2-Hydroxyethyl)benzoic acid 95%

Supplier: Ambeed
Warning

A101353-1G A101353-250MG A101353-5G
103795-926EA 198 USD
103795-926 103821-614 77282-706 77282-708 77282-710
4-(2-Hydroxyethyl)benzoic acid 95%
4-(2-Hydroxyethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.174 g/mol
Melting Pt: 122…129 °C
Storage Temperature: Ambient
MDL Number: MFCD02114379
CAS Number: 46112-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR