4-(2-Chlorothieno[3,2-d]pyrimidin-4-yl)morpholine 98%

Supplier: Ambeed
Warning

A199351-100MG A199351-250MG A199351-5G A199351-1G
103817-098EA 80 USD
103817-098 76785-248 76785-250 76785-252 77302-618
4-(2-Chlorothieno[3,2-d]pyrimidin-4-yl)morpholine 98%
4-(2-Chlorothieno[3,2-d]pyrimidin-4-yl)morpholine

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClN₃OS
MW: 255.7200 g/mol
Boiling Pt: 393.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09909656
CAS Number: 16234-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR