4-(2-Bromoethyl)phenol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxyphenethyl bromide

A297878-10G A297878-250MG A297878-1G A297878-25G A297878-5G A297878-100G
103814-154EA 6 USD
103814-154 103835-298 76693-352 76693-354 77305-720 77305-722 77305-724 77601-334
4-(2-Bromoethyl)phenol 98%
4-(2-Bromoethyl)phenol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.06 g/mol
Melting Pt: 88…92 °C
Flash Pt: 121.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00129679
CAS Number: 14140-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR