2-(4-Morpholine)ethylbromide hydrobromide 98%

Supplier: Ambeed
Warning

A116970-100G A116970-10G A116970-250MG A116970-1G A116970-25G
103798-810EA 7 USD
103798-810 103824-682 76870-044 76870-046 76870-048 77311-474 77885-071
2-(4-Morpholine)ethylbromide hydrobromide 98%
2-(4-Morpholine)ethylbromide hydrobromide

Precaución: for Research Use Only

Formula: C₆H₁₃Br₂NO
Melting Pt: 222…224 °C
Storage Temperature: Ambient
MDL Number: MFCD00463351
CAS Number: 42802-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR