4-(2-Bromoethyl)benzoic acid methyl ester 97%

Supplier: Ambeed
Warning

Synonyms: 4-(2-Bromoethyl)benzoic acid

A243526-1G A243526-25G A243526-5G A243526-10G A243526-250MG
103811-520EA 30 USD
103811-520 103851-342 77090-536 77090-538 77090-540 77288-786 77288-788
4-(2-Bromoethyl)benzoic acid methyl ester 97%
4-(2-Bromoethyl)benzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.0706 g/mol
Boiling Pt: 332.5 °C (760 mm Hg)
Melting Pt: 207…211 °C
Flash Pt: 154.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00016541
CAS Number: 52062-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR