4-(2-Bromoethyl)-1,2-difluorobenzene 97%

Supplier: Ambeed
Danger

A247497-250MG A247497-1G A247497-5G
103811-774EA 84 USD
103811-774 103851-610 76826-874 77090-948 77304-416
4-(2-Bromoethyl)-1,2-difluorobenzene 97%
4-(2-Bromoethyl)-1,2-difluorobenzene

Precaución: for Research Use Only

Formula: C₈H₇BrF₂
MW: 221.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12025017
CAS Number: 175018-77-6
UN: 1760
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR