4-(2-Aminoethyl)tetrahydropyran 97%

Supplier: Ambeed
Warning

Synonyms: 4-Aminoethyl-tetrahydropyrane, 4-(2-Aminoethyl)tetrahydro-2H-pyran

A220360-1G A220360-250MG A220360-100MG
103818-852EA 167 USD
103818-852 103849-734 77088-322 77112-724 77612-148
4-(2-Aminoethyl)tetrahydropyran 97%
4-(2-Aminoethyl)tetrahydropyran

Precaución: for Research Use Only

Formula: C₇H₁₅NO
MW: 129.2001 g/mol
Flash Pt: 74.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179433
CAS Number: 65412-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR