4-(2,4-Difluorobenzoyl)piperidine hydrochloride 98%

Supplier: Ambeed
Warning

A180671-25G A180671-1G A180671-5G
103807-550EA 9 USD
103807-550 103845-910 77083-412 77301-684 77301-686
4-(2,4-Difluorobenzoyl)piperidine hydrochloride 98%
4-(2,4-Difluorobenzoyl)piperidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₃F₂NO
Storage Temperature: Ambient
MDL Number: MFCD01313310
CAS Number: 106266-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR