4-(2,2,2-Trifluoroethyl)aniline 95%

Supplier: Ambeed
Warning

A377547-100MG A377547-250MG
103826-610EA 181 USD
103826-610 103837-640 76876-614 76876-616
4-(2,2,2-Trifluoroethyl)aniline 95%
4-(2,2,2-Trifluoroethyl)aniline

Precaución: for Research Use Only

Formula: C₈H₈F₃N
MW: 175.15 g/mol
Melting Pt: 45…46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04972985
CAS Number: 131395-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR