4-(2,2,2-Trifluoroethoxy)benzoic acid 97%

Supplier: Ambeed
Warning

A104751-100MG A104751-10G A104751-250MG A104751-5G
103796-774EA 37 USD
103796-774 103822-498 77148-394 77148-396 77148-398 77309-358
4-(2,2,2-Trifluoroethoxy)benzoic acid 97%
4-(2,2,2-Trifluoroethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₃
MW: 220.145 g/mol
Boiling Pt: 272.0±40.0 °C (760 mm Hg)
Flash Pt: 118.3 °C
Storage Temperature: Ambient
MDL Number: MFCD05669500
CAS Number: 27914-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR