4-(2,2,2-Trifluoroacetamido)butanoic acid 95%

Supplier: Ambeed
Warning

A296272-1G A296272-250MG
103814-088EA 57 USD
103814-088 103835-228 76837-894 76875-590
4-(2,2,2-Trifluoroacetamido)butanoic acid 95%
4-(2,2,2-Trifluoroacetamido)butanoic acid

Precaución: for Research Use Only

Formula: C₆H₈F₃NO₃
MW: 199.128 g/mol
Boiling Pt: 328.3 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00130210
CAS Number: 50632-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR