4-(2-Pyrrolidin-1-ylethoxy)phenylamine 95%

Supplier: Ambeed
Warning

Synonyms: 4-(2-(Pyrrolidin-1-yl)ethoxy)aniline

A144803-100MG A144803-250MG
103803-448EA 175 USD
103803-448 77285-888 77285-890
4-(2-Pyrrolidin-1-ylethoxy)phenylamine 95%
4-(2-Pyrrolidin-1-ylethoxy)phenylamine

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O
MW: 206.29 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07790198
CAS Number: 50609-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR