1-(2-Morpholinoethyl)-1H-pyrazole-4-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 4-(2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)ethyl)morpholine

A130876-250MG A130876-1G A130876-5G
103801-488EA 33 USD
103801-488 77027-050 77313-958 77313-960
1-(2-Morpholinoethyl)-1H-pyrazole-4-boronic acid pinacol ester 98%
1-(2-Morpholinoethyl)-1H-pyrazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₆BN₃O₃
Melting Pt: 89…94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08062342
CAS Number: 864754-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR