4-(2-((tert-Butoxycarbonyl)amino)ethyl)benzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-(2-Boc-aminoethyl)benzoic acid

A119439-10G A119439-250MG A119439-1G A119439-25G
103799-766EA 22 USD
103799-766 77009-358 77009-360 77297-682 77683-072
4-(2-((tert-Butoxycarbonyl)amino)ethyl)benzoic acid 95%
4-(2-((tert-Butoxycarbonyl)amino)ethyl)benzoic acid

Precaución: For research use only.

Formula: C₁₄H₁₉NO₄
MW: 265.3050 g/mol
Boiling Pt: 440.5 °C (760 mm Hg)
Flash Pt: 220.235 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07371516
CAS Number: 132690-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR