4-(1-Methylpiperidin-4-yl)aniline 95%
Supplier: Ambeed
|
Warning
|
A269838-1G
A269838-5G
A269838-100MG
A269838-10G
A269838-250MG
103812-978EA
39
USD
103812-978
103852-884
77092-652
77092-654
77166-398
77166-400
77166-402
4-(1-Methylpiperidin-4-yl)aniline 95%
4-(1-Methylpiperidin-4-yl)aniline
Precaución: for Research Use Only
Formula:
C₁₂H₁₈N₂ Flash Pt: 130.079 °C Storage Temperature: Ambient |
MDL Number:
MFCD06804550 CAS Number: 454482-12-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...