4-(1-Methyl-1H-pyrazol-4-yl)benzoic acid 97%
Supplier: Ambeed
|
Warning
|
A144378-100MG
A144378-250MG
A144378-5G
A144378-1G
103803-396EA
275
USD
103803-396
77272-622
77272-624
77272-626
77299-374
4-(1-Methyl-1H-pyrazol-4-yl)benzoic acid 97%
4-(1-Methyl-1H-pyrazol-4-yl)benzoic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₀N₂O₂ MW: 202.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11932712 CAS Number: 199678-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...