4-(1H-Pyrazol-4-yl)aniline 97%
Supplier: Ambeed
|
Warning
|
A105393-100MG
A105393-250MG
A105393-1G
A105393-5G
103796-886EA
67
USD
103796-886
103822-614
76809-556
76809-558
77283-360
77309-486
4-(1H-Pyrazol-4-yl)aniline 97%
4-(1H-Pyrazol-4-yl)aniline
Precaución: for Research Use Only
Formula:
C₉H₉N₃ MW: 159.1900 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11932600 CAS Number: 114474-28-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...