4-(1H-Benzimidazol-2-yl)aniline 95%

Supplier: Ambeed
Warning

A240163-1G A240163-250MG A240163-5G
103811-330EA 70 USD
103811-330 103851-128 77090-278 77288-712 77304-184
4-(1H-Benzimidazol-2-yl)aniline 95%
4-(1H-Benzimidazol-2-yl)aniline

Precaución: for Research Use Only

Formula: C₁₃H₁₁N₃
Boiling Pt: 466.3 °C (760 mm Hg)
Flash Pt: 266.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00454395
CAS Number: 2963-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR