Compound A 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-((4-acetophenyl)-2-chloro-N-methyl)ethylammonium chloride, 4-(1-Chloro-2-(methylamino)ethyl)phenyl acetate hydrochloride
A227582-100MG
A227582-250MG
A227582-1G
103819-328EA
49
USD
103819-328
103850-234
77000-100
77000-102
77303-804
Compound A 97%
Compound A
Precaución: for Research Use Only
Formula:
C₁₁H₁₄ClNO₂·HCl Boiling Pt: 324.6 °C (760 mm Hg) Flash Pt: 150.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD16883076 CAS Number: 14593-25-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...