4-(1-Aminocyclopropyl)benzonitrile hydrochloride 97%

Supplier: Ambeed
Warning

A391285-100MG A391285-250MG
103826-882EA 192 USD
103826-882 77016-190 77016-192
4-(1-Aminocyclopropyl)benzonitrile hydrochloride 97%
4-(1-Aminocyclopropyl)benzonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD18207017
CAS Number: 1369512-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR