4-(1,3-Oxazol-5-yl)aniline 97%

Supplier: Ambeed
Warning

A132786-250MG A132786-1G
103801-780EA 39 USD
103801-780 77285-378 77314-364
4-(1,3-Oxazol-5-yl)aniline 97%
4-(1,3-Oxazol-5-yl)aniline

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MW: 160.1700 g/mol
Boiling Pt: 318.7 °C (760 mm Hg)
Melting Pt: 148 °C
Flash Pt: 146.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00052192
CAS Number: 1008-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR