4-[(Trimethylsilyl)ethynyl]benzaldehyde 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-((Trimethylsilyl)ethynyl)benzaldehyde, 4-(Trimethylsilylethynyl)benzaldehyde
A281335-25G
A281335-5G
A281335-10G
A281335-1G
103813-546EA
7
USD
103813-546
103853-496
76837-194
76837-196
77141-182
77305-388
4-[(Trimethylsilyl)ethynyl]benzaldehyde 95%
4-[(Trimethylsilyl)ethynyl]benzaldehyde
Precaución: for Research Use Only
Formula:
C₁₂H₁₄OSi Boiling Pt: 262.1 °C (760 mm Hg) Melting Pt: 66…70 °C Flash Pt: 112.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093765 CAS Number: 77123-57-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...