4-[(Trifluoromethyl)thio]aniline 98%

Supplier: Ambeed
Warning

Synonyms: 4-((Trifluoromethyl)thio)aniline

A203208-1G A203208-25G A203208-5G A203208-10G A203208-500G
103817-446EA 5 USD
103817-446 103848-100 77086-222 77086-224 77086-226 77287-728 77287-730
4-[(Trifluoromethyl)thio]aniline 98%
4-[(Trifluoromethyl)thio]aniline

Precaución: for Research Use Only

Formula: C₇H₆F₃NS
MW: 193.19 g/mol
Boiling Pt: 183 °C (760 mm Hg)
Flash Pt: 64.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00040926
CAS Number: 372-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR