4-N-Boc-amino-4-carboxytetrahydropyran 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-((tert-Butoxycarbonyl)amino)tetrahydro-2H-pyran-4-carboxylic acid
A245736-1G
A245736-25G
A245736-5G
A245736-100MG
A245736-10G
A245736-250MG
103811-664EA
31
USD
103811-664
103851-496
77090-762
77090-764
77090-766
77288-870
77288-872
77288-874
4-N-Boc-amino-4-carboxytetrahydropyran 97%
4-N-Boc-amino-4-carboxytetrahydropyran
Precaución: for Research Use Only
Formula:
C₁₁H₁₉NO₅ MW: 245.275 g/mol Boiling Pt: 407.9 °C (760 mm Hg) Flash Pt: 200.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02683136 CAS Number: 172843-97-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...