4-((tert-Butoxycarbonyl)amino)piperidine-4-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A103006-100MG
A103006-250MG
A103006-1G
103796-464EA
15
USD
103796-464
103822-166
77107-310
77107-312
77309-032
4-((tert-Butoxycarbonyl)amino)piperidine-4-carboxylic acid 95%
4-((tert-Butoxycarbonyl)amino)piperidine-4-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₁H₂₀N₂O₄ MW: 244.28 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03790944 CAS Number: 252720-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...