4-((tert-Butoxycarbonyl)amino)bicyclo[2.2.2]octane-1-carboxylic acid 97%

Supplier: Ambeed
Warning

A144489-100MG A144489-5G A144489-1G A144489-25G
103803-412EA 111 USD
103803-412 76823-588 76823-590 77299-388 77683-241
4-((tert-Butoxycarbonyl)amino)bicyclo[2.2.2]octane-1-carboxylic acid 97%
4-((tert-Butoxycarbonyl)amino)bicyclo[2.2.2]octane-1-carboxylic acid

Precaución: For research use only.

Formula: C₁₄H₂₃NO₄
MW: 269.337 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22577789
CAS Number: 863304-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR