4-((tert-Butoxycarbonyl)(methyl)amino)butanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Boc-N-methyl-gamma-aminobutyric acid

A352699-100MG A352699-250MG A352699-25G
103826-074EA 52 USD
103826-074 103837-076 76876-308 76876-310 77672-774
4-((tert-Butoxycarbonyl)(methyl)amino)butanoic acid 98%
4-((tert-Butoxycarbonyl)(methyl)amino)butanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
MW: 217.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09831984
CAS Number: 94994-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR