[4-[(Dimethylamino)methyl]phenyl]boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-((Dimethylamino)methyl)phenylboronic acid

A177946-100MG A177946-10G A177946-250MG A177946-1G
103807-280EA 24 USD
103807-280 103845-620 76974-324 76974-326 76974-328 77083-016
[4-[(Dimethylamino)methyl]phenyl]boronic acid 97%
[4-[(Dimethylamino)methyl]phenyl]boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₄BNO₂
MW: 179.024 g/mol
Boiling Pt: 297.4 °C (760 mm Hg)
Flash Pt: 133.6 °C
Storage Temperature: Freezer
MDL Number: MFCD01319001
CAS Number: 70799-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR