4-Dimethylaminomethylbenzoic acid hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4-((Dimethylamino)methyl)benzoic acid hydrochloride

A126406-100MG A126406-250MG
103800-806EA 111 USD
103800-806 77161-640 77161-642
4-Dimethylaminomethylbenzoic acid hydrochloride 97%
4-Dimethylaminomethylbenzoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClNO₂
MW: 215.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09997604
CAS Number: 17847-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR