4-[(Benzyloxy)carbonyl]-1-(tert-butoxycarbonyl)piperazine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-((Benzyloxy)carbonyl)-1-(tert-butoxycarbonyl)piperazine-2-carboxylic acid, Piperazine-2-carboxylic acid, N1-BOC N4-CBZ protected

A206828-250MG A206828-1G A206828-25G A206828-5G
103817-734EA 43 USD
103817-734 103848-414 76999-532 77086-670 77086-672 77303-010
4-[(Benzyloxy)carbonyl]-1-(tert-butoxycarbonyl)piperazine-2-carboxylic acid 95%
4-[(Benzyloxy)carbonyl]-1-(tert-butoxycarbonyl)piperazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₈H₂₄N₂O₆
MW: 364.4 g/mol
Boiling Pt: 518.9 °C (760 mm Hg)
Flash Pt: 267.6 °C
Storage Temperature: Ambient
MDL Number: MFCD02683055
CAS Number: 149057-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR