4-((6-(Acryloyloxy)hexyl)oxy)benzoic acid 97%
Supplier: Ambeed
|
Warning
|
A110759-10G
A110759-1G
A110759-250MG
A110759-25G
A110759-5G
103797-720EA
9
USD
103797-720
103823-530
77284-034
77284-036
77284-038
77284-040
77310-330
4-((6-(Acryloyloxy)hexyl)oxy)benzoic acid 97%
4-((6-(Acryloyloxy)hexyl)oxy)benzoic acid
Precaución: for Research Use Only
Formula:
C₁₆H₂₀O₅ MW: 292.33 g/mol Boiling Pt: 446.9 °C (760 mm Hg) Flash Pt: 160.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01166387 CAS Number: 83883-26-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...