4-[(4-Methylpiperazin-1-yl)methyl]-3-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Warning

A568431-1G A568431-25G A568431-10G A568431-250MG A568431-5G
103828-792EA 17 USD
103828-792 103839-934 76844-058 76844-060 76878-686 76878-688 77307-838
4-[(4-Methylpiperazin-1-yl)methyl]-3-(trifluoromethyl)aniline 98%
4-[(4-Methylpiperazin-1-yl)methyl]-3-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₁₃H₁₈F₃N₃
Melting Pt: 116…120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09909924
CAS Number: 694499-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR