4-[(4-Isopropoxyphenyl)sulfonyl]phenol 97%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxyphenyl 4-Isopropoxyphenyl sulphone, 4-((4-Isopropoxyphenyl)sulphonyl)phenol

A312018-100G A312018-10G A312018-1G A312018-25G A312018-5G
103814-688EA 8 USD
103814-688 103835-878 76875-768 76875-770 77306-062 77306-064 77306-066
4-[(4-Isopropoxyphenyl)sulfonyl]phenol 97%
4-[(4-Isopropoxyphenyl)sulfonyl]phenol

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₄S
MW: 292.35 g/mol
Boiling Pt: 478.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01861244
CAS Number: 95235-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR