4-((1-Methylpiperidin-4-yl)oxy)aniline 97%

Supplier: Ambeed
Warning

A884540-5G A884540-100MG A884540-250MG A884540-1G
103841-322EA 570 USD
103841-322 76845-590 77019-848 77019-850 77308-488
4-((1-Methylpiperidin-4-yl)oxy)aniline 97%
4-((1-Methylpiperidin-4-yl)oxy)aniline

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O
MW: 206.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04970985
CAS Number: 358789-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR