4-[(tert-Butoxycarbonylamino)methyl]benzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-(((tert-Butoxycarbonyl)amino)methyl)benzoic acid, 4-[(Boc-amino)methyl]benzoic Acid, Boc-4-Amb-OH

A178848-1G A178848-25G A178848-5G A178848-100G A178848-10G A178848-500G
103807-376EA 10 USD
103807-376 103845-716 77083-160 77083-162 77083-164 77163-796 77163-798 77163-800
4-[(tert-Butoxycarbonylamino)methyl]benzoic acid 97%
4-[(tert-Butoxycarbonylamino)methyl]benzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₄
MW: 251.28 g/mol
Boiling Pt: 427.8 °C (760 mm Hg)
Melting Pt: 164…168 °C
Flash Pt: 212.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00228182
CAS Number: 33233-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR