4-{[(2-Amino-4-hydroxypteridin-6-yl)methyl]amino}benzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Pteroic acid, 4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzoic acid, Folic Acid Impurity D

A159632-100MG A159632-10G A159632-250MG A159632-1G A159632-25G A159632-5G
103805-270EA 18 USD
103805-270 103843-492 76720-064 76720-066 76720-068 77319-382 77319-384 77319-386
4-{[(2-Amino-4-hydroxypteridin-6-yl)methyl]amino}benzoic acid 95%
4-{[(2-Amino-4-hydroxypteridin-6-yl)methyl]amino}benzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₆O₃
MW: 312.283 g/mol
Boiling Pt: 659.8 °C (760 mm Hg)
Flash Pt: 352.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075823
CAS Number: 119-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR