Fmoc-4-amino-tetrahydropyran-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A162624-10G A162624-250MG A162624-1G A162624-5G
103805-642EA 18 USD
103805-642 103843-892 77286-498 77286-500 77319-954 77319-956
Fmoc-4-amino-tetrahydropyran-4-carboxylic acid 95%
Fmoc-4-amino-tetrahydropyran-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₅
Boiling Pt: 609.9 °C (760 mm Hg)
Flash Pt: 322.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683137
CAS Number: 285996-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR